Would you like to log in again?
Separating a Mixture by Physical Properties Kit
Supplier: Avantor
WARD470039-664
470039-664EA 128.69 USD InStock
470039-664
Separating a Mixture by Physical Properties Kit
Educational Classroom Kits and Activities
Physically Separate Different Substances
  • Two Levels of Instruction Included
  • Determine properties of different substances and collect data
Students identify physical properties of substances and then determine methods that can separate a mixture made of those substances. Methods include separation by properties including particle size, density, solubility, and magnetism. Directions for two levels of instruction are included. The elementary level activity stresses identification properties, use of tools, and using collected data appropriately in designing an experiment. The advanced level activity includes some mathematical analysis for understanding physical properties of matter. one 45-minute class period is required to complete the activity. Includes Instruction manual. A balance is required but not included.
Includes:
Magnetic wants
Strainers
Funnels
Aluminum pie plates
Filter Paper
Plastic Spoons
Pop beads
Salt
Sand
Steel nuts
Pebbles
Plastic Cups
Plastic Containers
Instructions
Required but Not Provided: Balance, Beakers, Water, Containers, Paper Towels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty