Would you like to log in again?
Arthropod Classification Kit
Supplier: Avantor
WARD470039-702
470039-702EA 80.99 USD InStock
470039-702
Arthropod Classification Kit
Educational Classroom Kits and Activities
From introductory level to advanced study, this kit provides engaging lessons on insect classification and arthropod characteristics.
  • Lessons on insects and arthropods
  • Two levels of instruction included
Two separate levels of instruction are provided for each step of this activity. Beginning students will use a table to classify plastic models of insects, while advanced level students use a dichotomous key to identify models as insects, arachnids, crustaceans, and centipedes and millipedes. Additional advanced level study asks students to construct their own dichotomous key for arthropod classification. Developed by Kimberly Rogers. Includes instructions for two levels of activity and materials for 12 student groups. Grades 4–10.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty