Would you like to log in again?
Color Mixing Apparatus
Supplier: United Scientific Supplies
CMA001
470039-942EA 165.35 USD InStock
470039-942
Color Mixing Apparatus
Educational Materials Physics Educational Materials Optics Learning Activities
Illuminate the variances between additive and subtractive color mixing
  • Independent control for each color
  • Investigate additive and subtractive color mixing
A compact and economical Color Mixing Apparatus. This updated version of the classic three color light projector uses three low voltage, high power LED's to project easily-seen red, green, and blue light onto a white screen. The projector mounting allows the beam angles to be adjusted to vary the overlap of the three colored images. The intensity of each LED can be adjusted separately by a potentiometer to give a convincing white light or various shades of mixed colors. Colored shadows can also be explored in a darkened room. The set includes a wall mount power supply and activity.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty