Would you like to log in again?
Friction on an Inclined Plane
Supplier: United Scientific Supplies
INPW02
470039-944EA 95.15 USD InStock
470039-944
Friction on an Inclined Plane
Educational Materials Physics Educational Materials Mechanics Learning Activities
This set demonstrates a wide variety of physical science topics including the resolution of forces on an inclined plane, kinetic and potential energies, uniform and accelerated motions, and the friction between two surfaces.
  • Inclined Board Can Be Set to a Range of Angles
  • For Studying Concepts Related to Friction
  • Instructions Included
  • Board Length: 60 cm
  • Board Width: 10 cm
Students can explore and determine the coefficients of static and sliding friction and also verify normal and frictional components of the force exerted by the plane.
The 10 x 60 cm wooden inclined board can be set at any angle between zero and 45°, and angle measurements can be made directly through the two scales on the included protractor.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty