Would you like to log in again?
Electromagnet Kit
Supplier: United Scientific Supplies
EMKIT2
470039-946EA 44.55 USD InStock
470039-946
Electromagnet Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Reliable kit in a quality carrying case!
  • Build bar and U shaped electromagnets
  • The kit includes a 5 x 5 x 1 1/2" plastic case
You will receive two coils on plastic spools, a U shaped iron core, a straight iron core, two magnetic needles with stands, and a pair of wires. Students can use these materials to build bar and U shaped electromagnets with a 6 V battery or DC power supply, and determine the magnitude and polarity of electromagnetic forces. Includes instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty