Would you like to log in again?
Create-a-Circuit Kit
Supplier: United Scientific Supplies
CCK001
470039-950EA 31.6 USD InStock
470039-950
Create-a-Circuit Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Create-a-Circuit kit gives teachers a hands-on system for introducing students to circuitry.
  • Instructions Included
  • Ideal Introduction to Circuitry
  • Hands-On Activity
  • Kit Contains All Items Required to Make a Series Circuit
The kit features a "D" cell battery holder, knife switch, two miniature light bulbs, two lamp holders, and four connector cords.
It has everything needed to build a series circuit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty