Would you like to log in again?
Mystery Crooked Cell Activity
Supplier: Edvotek
S-53
470039-968EA 105.3 USD InStock
470039-968
Mystery Crooked Cell Activity
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Simulate Genetic Diagnosis of Sickle Cell Anemia
  • Real world relevance
  • Time required: 45 minutes
This simple lab demonstrates detection of the mutation that causes Sickle Cell Anemia. In this simulation, your students will use electrophoresis to separate dyes that represent patient samples and controls
Kit includes: instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, microtipped transfer pipettes.
Required, but not included: electrophoresis apparatus, power supply, microwave or hot plate.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty