Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylene

Supplier: ALDON CORP SE
XX0020-500ML XX0020-500MLR
470045-580EA 25.2 USD InStock
470045-580 470303-058
Xylene
Chemicals for Science Education
CAS Number: 1330-20-7
Formula Weight: 106.17
Formula: C6H4(CH3)2
Density (g/mL): 0.87
Boiling Point (°C): 140
Shelf Life (months): 36
Storage: Red

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty