Would you like to log in again?
Variable Speed Wave Generator
Supplier: THE SCIENCE SOURCE SE
15440
470045-910EA 111 USD InStock
470045-910
Variable Speed Wave Generator
Models Stream Models
The Variable Speed Wave Generator is a new concept for generating waves in your stream table. The paddle enters the water, forms the wave, then is withdrawn to eliminate back waves.
The frequency of waves can be varied by increasing or reducing the voltage to the unit. The wave generator is supplied with an eight inch rod for assembly to an upright support making adjustment of wave direction and depth of paddle easy. This unit requires a variable low voltage power supply, 0 to 6 V DC, 0 to 1 amp or a 1.5V #6 dry cell and rheostat.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty