Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

500 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer

Supplier: INTERNATIONAL VIDEO & ELECTRONICS
DS-500
470061-802EA 102.3 USD InStock
470061-802
500 Watt Type 3 Voltage Converter, Step Up and Step Down Voltage Transformer
Power Supplies
The International DS-500 Diamond series 500 W type 3 V converters are 220 Electronics best voltage transformers. They are thoroughly tested to the highest workmanship and material standards and hand soldering insures to meet U.S Military specifications.

  • USA grounded male plug
  • Extra adapter included to use with 220 V outlets
  • Converts 110 V to 220 V and vice versa
  • Extra durable lifetime coil are second to none
  • Works anywhere in the world
  • Heavy duty for continuous use
  • Extra durable lifetime coil is included
  • Recommended for high value and/or sensitive items

Ideal when visiting or moving overseas.

Weight: 8 lbs. (3.6 kg)
Dimensions: 4.75x6x4" (12.1x15.2x10.2 cm)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty