Would you like to log in again?
Eisco® Cardiac Muscle
Supplier: EISCO SCIENTIFIC LLC
AM0082
470094-024EA 222.5 USD InStock
470094-024
Eisco® Cardiac Muscle
Models Human Biology Models
Examine the structures within the cardiac muscle fiber with this comprehensive model.
  • Students Observe Cardiac Muscle Fiber
  • Model Shows Damage from High Blood Pressure
  • Removable Organs
  • Model Represents the Cell Membrane, Sarcomere Myofibrils, and More
The cell membrane, sarcomere myofibrils, and sarcoplasmic reticulum are all represented.
The organs are removable for closer study.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty