Would you like to log in again?
Wacky Hall Walker
Supplier: EDUCATIONAL INNOVATIONS, INC.
WHW-100
470095-234EA 52.9 USD InStock
470095-234
Wacky Hall Walker
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Energy Transfer Demonstration
  • Explain the difference between kinetic and potential energy
  • Conduct experiments using the Wacky Hall Walker
  • Apply the law of conservation of energy as it relates to the Wacky Hall Walker
  • Make predictions based upon collected data
  • Measure distances in metric units
  • Construct tables and graphs to display data
The law of conservation of energy states that the amount of energy put into a system is equal to the amount of energy leaving the system. The elastic energy stored in the twisted rubber band of the Wacky Hall Walker (WHW) is called potential energy. The energy of motion released as the WHW moves is called kinetic energy. The number of twists applied to the rubber band mechanism is proportional to the distance the WHW travels. The motion of the WHW is also influenced by the size and mass of the WHW body, size of the rubber bands used inside the container, type of surface the WHW travels over, size of the pencil, and whether the eraser end of the pencil is dragging along the floor or up in the air.
Ordering information: Materials include Clear cylinder, 2 plastic end caps with a hole in the center, small round bead, #117 rubber band, metal washer, pencil, paper clip, masking tape, and measuring tape.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty