Would you like to log in again?
TeacherGeek Project Cylinder - Hydraulic or Pneumatic
Supplier: TeacherGeek
1823-52 1823-53
470096-122PK 48.75 USD InStock
470096-122 470096-124
TeacherGeek Project Cylinder - Hydraulic or Pneumatic
Educational Materials Physics Educational Materials TeacherGeek System Components
Bulk syringes to expand your TeacherGeek collection
TeacherGeek Project Cylinders are the best solution for school and maker hydraulic/pneumatic projects. They come in two sizes: 4.5ml and 14ml.
What make them so great?
Holes, pins and a flange for mounting to your projects
Screws to keep the plunger and tubing from coming out (keeping the water in your project)
Volume and distance graduations on the piston
Extra-long travel (the piston moves much further than on a syringe)
TeacherGeek Cylinders are not syringes. Their tip/port will not accept needles. This means that they are not regulated as a medical device (like syringes), and can be used in public school classrooms without issue.
Connect cylinders together using 1/8in vinyl tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty