Would you like to log in again?
Streak Plates
Supplier: Shiv Dial Sud & Sons
Avg. Ratings: 5.0 (3 Reviews)
REVIEWS (3)
Total Ratings: 3
Avg. Ratings: out of 5
Sort all by:
WARD470097-470 WARD470097-452
470097-470PK 11.55 USD InStock
470097-470 470097-452
Streak Plates
Staining Tiles
Streak color helps identify minerals.
  • Plates Sized for Field and Lab Testing
  • Unglazed Porcelain
  • Black Plates Work with Light Minerals
These unglazed porcelain plates are sized for field and lab testing. Rubbing the specimen across the plate reveals its streak color.
Black plates work with light minerals.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (3)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty