Would you like to log in again?
Characteristics of Matter Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
1-114
470100-890EA 242 USD InStock
470100-890
Characteristics of Matter Lab Activity
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Determine the Many Properties of Matter
  • Meets national standards for grades 6-9
  • Combine scientific inquiry with structured assessments
Ten experiments in which students differentiate between physical and chemical properties and explore the characteristics of matter; color, odor, solubility, shape, size, texture, density, alkalinity, acidity, states of matter, boiling point, and melting point. Includes graphing of melting and boiling points. Students observe chemical properties of five solutions by observing their reactions with other solutions. From these reactions, students then determine the identity of two unknown solutions. Includes reproducible student data sheets, assessment questions, and answer keys. Required, but not included: goggles, balance, ice, test tubes, beaker or clear cup, 10 mL graduated cylinder, and test tube racks.
Includes:
Instructor Manual
Spot Plates
Wood Stirrers
Thermometers
Plastic Bottles with Droppers & Caps
Blue Litmus
Red Litmus
Pipettes
Ammonia Solution
Baking Soda Solution
Calcium Chloride Solution
Vinegar
Washing Soda Solution
Calcium Chloride Solution
Calcium Carbonate
Sodium Chloride
Sand
Sucrose
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty