Would you like to log in again?
3-D Water Kit©
Supplier: 3-D Molecular Designs
CSWK-01 CSWK-06
470328-170CS 866.5 USD InStock
470328-170 470100-916
3-D Water Kit©
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
A fun tool to help teach water concepts.
  • Ideal for grades 5-12
  • Embedded magnets in atoms reflect the correct positive or negative charges
  • Students experience molecular attraction or repulsion hands-on
Your students will use these magnetic water molecules to discover hydrogen bonding, make ice, dissolve salt, evaporate water, explore transpiration, create ethanol, and much more. Water Kits© enhance learning through hand-on activity and facilitate more in depth concept conversations. Your students can feel the various strengths of hydrogen, covalent and ionic bonds. They can explore the water cycle – even make ice. They also can show the differences between molecular adhesion, cohesion and capillary action and demonstrate surface tension, evaporation, condensation and solubility.
Materials Included (per cup):
1 Plastic cup
12 Water molecules
1 Sodium
1 Chloride
1 Ethane molecule
1 Hydroxyl group
1 six-cup tray (case of 8 only)
Ordering information: Assembly required. Includes assembly instructions and lesson resources.
Caution: WARNING: SMALL MAGNETS- This product contains small magnets. Swallowed magnets can stick together across intestines, causing serious infections and death. Seek immediate medical attention if magnets are swallowed or inhaled.
WARNING: CHOKING HAZARD - This product contains small parts and should be kept out of the reach of children under the age of 3, because the parts or the pieces may present a choking hazard to small children.
CAUTION: This is a science education product, not a toy. It is not intended for children under 8 years old.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty