Would you like to log in again?
Glow Chemistry Kit
Supplier: Avantor
WARD470101-046 GP16BRITEG GP16BRITEB GP16BRITER
470101-046KT 134.19 USD InStock
470101-046 470106-828 470106-826 470106-824
Glow Chemistry Kit
Educational Classroom Kits and Activities
Discover the Chemistry Behind Luminescence
  • Easy to use
  • For grades 11-12
  • Replacement light sticks available separately, in various colors
This demonstration lets you mix sodium hydroxide, dimethyl sulphoxide and luminol to produce a strong blue-green light. Even after the light fades, shaking the bottle will cause more of the luminol to react. Use the supplied light stick to teach students about practical applications of luminescence. Developed by Lynn Hesse.
Ordering information: Light sticks have a shelf life of 18 months.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty