Would you like to log in again?
Cellular Anatomy Notebook Chart
Supplier: PERMACHARTS
1550803778
470101-368EA 13.15 USD InStock
470101-368
Cellular Anatomy Notebook Chart
Charts
Introduce your students to cellular anatomy with this vivid, full-color, two-sided chart.
  • Laminated for durability
One side depicts the richly detailed, inner structure of a plant cell and organelles, while the other is a generalized animal cell with organelles. The result of extensive research from multiple sources, each component is clearly labeled. The animal cell side features plasma membrane, nuclear envelope, and cytoskeleton details. This quick reference chart is laminated for durability and protection against spills and other damage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty