Would you like to log in again?
Cancer Cell Detection Lab Activity Kit
Supplier: Edvotek
115
470101-432EA 138.7 USD InStock
470101-432
Cancer Cell Detection Lab Activity Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
Biotechnology-Based Diagnostic Tests
  • Connected to cancer genetics
  • Time required: 45 minutes
Immortality through uncontrolled cell division is a characteristic of cancer cells. The p53 gene is a tumor suppressor gene which prevents this. Mutations in this gene are present in more than 50% of cancers. Testing people for mutations in their p53 gene can indicate an increased risk in developing cancer. These tests raise intriguing ethical questions for both the individual tested and the family of an individual who chooses to be tested. In this experiment, students determine a pedigree for a family suspected to be carriers of mutations in their p53 genes. A DNA test indicates their likelihood of developing cancer.
Kit includes: instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, calibrated pipette, microtipped transfer pipettes, InstaStain® Blue and FlashBlue™ stain.
Required, but not included: electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, visualization system.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty