Would you like to log in again?
A-Frame Base with Rod
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ABSE12 ABSE12-SPL
470101-474EA 52.5 USD InStock
470101-474 470101-476
A-Frame Base with Rod
Support Stands
These cast-iron bases have three-point stability on nearly any surface.
  • Rod and base set consisting of an A-shaped retort base
  • Baked enamel finish
  • Stackable even with rod attached
The retort stand is made of corrosion resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10×1.5 mm metric thread. Heavy weight construction provides superb stability for basic experiments and setups.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty