Would you like to log in again?
Urine Test Strips
Supplier: LW SCIENTIFIC SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
URS-05PR-7777 URS-10PR-7777
470210-690PK 64.1 USD InStock
470210-690 470101-700
Urine Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
Easy-to-use test strips check for up to 10 parameters.
  • Easy to Use
5 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone
10 Parameter Urine Reagent Strips: Glucose, Protein, pH, Blood, Ketone, bilirubin, nitrite, urobilinogen, specific gravity, and leukocytes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty