Would you like to log in again?
Lab-Aids® Genetic Equilibrium And Natural Selection Kit
Supplier: LAB-AIDS INC (KITS) SE
76
470101-702EA 129 USD InStock
470101-702
Lab-Aids® Genetic Equilibrium And Natural Selection Kit
Educational Classroom Kits and Activities
Examine the impacts of genetic equilibrium and selection on evolution
  • Understand the principles of genetic equilibrium and natural selection as a process of evolution
  • Collect data from a simulated genetic process when natural selection is operating on a population in genetic equilibrium
  • Use data in a Hardy-Weinberg equilibrium
  • Reusable
  • Time required: One 50 minutes class period
In this lab, students study and examine genetic equilibrium and natural selection as processes of evolution. Simulations are used to develop insights about the actual genetic process involved when natural selection is operating on a population in equilibrium. Students evaluate data gathered from these simulations according to the Hardy-Weinberg law, leading to a better understanding of the staggering complexity of natural selection in nature.
Included:
Teacher’s guide
Student worksheets and guides
Shakers
Alleles, plastic, dark-colored
Alleles, plastic, light-colored
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty