Would you like to log in again?
Density of Hot and Cold Water Kit
Supplier: Avantor
WARD470101-810
470101-810EA 187.99 USD InStock
470101-810
Density of Hot and Cold Water Kit
Educational Classroom Kits and Activities
What happens when you place a separator sheet between a column of red-colored warm water on top of blue-colroed warm water and slowly slide the separator out? What happens if the cold water is on the top and the warm water is on the bottom? Use this lab to find out the answers to these questions and determine if density is an intrinsic or extrinsic property of matter.
  • Grades 4 to 8
Claire Zurbuck developed this kit to teach about water currents, air currents, and convection. Includes materials for five student groups and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty