Would you like to log in again?
Reaction Types, Gas Collection & More Kit
Supplier: Avantor
WARD470102-266
470102-266KT 224.79 USD InStock
470102-266
Reaction Types, Gas Collection & More Kit
Educational Classroom Kits and Activities
Explore the Four Reaction Types
  • Students make hypotheses and determinations about experiments
  • Great for grades 10-12
All four reaction types are examined in this lab – synthesis, decomposition, single replacement, and double replacement. In two of the five activities, students learn the technique of collecting gas over water, then test the gasses to see what they are. They also have to determine which reaction type each part of the lab produces. Developed by Mary J. Gallup.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty