Would you like to log in again?
3B Scientific® Pulmonary Lobule And Blood Vessels
Supplier: AMERICAN 3B SCIENTIFIC SE
1008493
470103-204EA 484.6 USD InStock
470103-204
3B Scientific® Pulmonary Lobule And Blood Vessels
Models Human Biology Models
  • Magnetic Assembly, No Pins
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
The model shows an external pulmonary lobe with a magnification of 130x. The following are represented: Segmental bronchus and its terminal branches (bronchioles, Alveolus opened on the right side, Pulmonary vessels and their capillary networks, Branch of a bronchial artery, Pulmonary pleura +, Connective tissue septum on the left side, Single opened alveolus with surrounding capillary network with a magnification of approx. 1000x on the rear side.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty