Would you like to log in again?
Laerdal® Resusci Junior CPR Manikin
Supplier: AMERICAN 3B SCIENTIFIC SE
1005204 1005205
470103-348PK 149.4 USD InStock
470103-348 470119-772
Laerdal® Resusci Junior CPR Manikin
High Quality CPR Training On A Child Sized Manikin
  • Educationally effective by focusing on the performance level of key cpr skills and techniques for children
  • Enhanced student motivation through use of the optional skill guide indicator that allows real-time cpr performance feedback during training
  • Cost efficient through easy to use and maintain disposable airways
  • Realistic anatomical body and landmarks provide the essential features necessary to learn realistic child handling and quality child cpr skills
  • Logistically convenient transportation provided by a durable hard case
The Resusci Junior manikin offers realistic, high-quality child CPR education. It has full-body construction and use of an optional Skill Guide performance indicator. Students can quickly sharpen their skills in performing child CPR to high standards. Features: Natural obstruction of the airway allows students to learn the important technique of opening the airway so that in a real-life situation they know how to effectively administer air to the patient, Head tilt/chin lift and jaw thrust allows students to correctly practice airway maneuvers necessary when resuscitating a real child patient, Realistic resistance for chest compressions allows the students to experience the amount of pressure needed to perform proper chest compressions in a real-life situation, Anatomically correct landmarks and sternal notch allow the student to practice identification of all anatomical landmarks relevant to child CPR.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty