Would you like to log in again?
Laerdal® Nursing Kid Manikin
Supplier: AMERICAN 3B SCIENTIFIC SE
1005275 1005244 1005249
470103-392EA 394.99 USD InStock
470103-392 470103-376 470120-378
Laerdal® Nursing Kid Manikin
Models Human Biology Models
Clinical Training On A 6 Year Old Child Trainer
  • Highly mobile for use in multiple clinical settings and hospital transport
  • Efficient training care and management of a variety of both common and uncommon in-hospital pediatric patients including wound assessment and care, first aid and child abuse training
  • Durable, rugged and lifelike; made to withstand years of use
The clinical skills training manikin for in-hospital pediatric professionals. Nursing Kid is a training manikin realistically representing a six-year old child. He is designed for skill and scenario based training of the care and management of a variety of pediatric in-hospital patients. Training includes sound auscultation, IV cannulation, urinary catheterization, and general pediatric patient care. This child is ideal for clinical training in core pediatric in-hospital clinical skills. Features: Head with anatomical landmarks, trachea, and esophagus along with simulated lungs and stomach allow the practice of many procedures, including NG, OG, tracheal care and suctioning, and insertion, securing and care of endotracheal tube. Allows for the auscultation and diagnosis of normal and abnormal hear, breath and bowel sounds. Bilateral deltoid, bilateral thigh, gluteal intramuscular injections possible. Articulating IV arm allows for practice of IV cannulation, medication administration and site care and maintenance. Presents normal and abnormal heart, breath and bowel sounds for auscultation. Supplied with: Full-Body Pediatric Manikin, manikin lubricant, simulated blood, Hospital Gown, Carry Case, Directions for Use.
Ordering information: Does not include a SimPad Unit.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty