Would you like to log in again?
Acid Rain and Plant Growth Kit, 30 Students
Supplier: CAROLINA BIOL SUPPLY CO SE
W 653075
470103-596EA 204.05 USD InStock
470103-596
Acid Rain and Plant Growth Kit, 30 Students
Educational Materials Biology Educational Materials Plant Learning Activities
What is acid rain doing to your home town?
  • Test the impact of acid rain on different materials
  • Test local rainwater
  • Includes a teacher's manual
The effects of acid rain have emerged as a devastating result of air pollution.
So how severe is the problem in your area? This kit allows students to test the pH of rain, discover the effects of pH on plants and observe the effect of substrate on pH.
Includes: marigold seed, marble chips, granite chips, buffers pH3, test cups, hydrochloric acid (1N), sodium sulfite, petri dishes, germination blotters, 10oz plastic cups, pH test paper, azolitmin solution, sodium bicarbonate, self-sealing bags, student instructions and a teacher's manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty