Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Bell Jar without Stopper
Supplier: VARIETY GLASS INC
636-1/2B-1
470104-014EA 142.95 USD InStock
470104-014
Bell Jar without Stopper
Jars Bell Jars
Made from fine, heavy molded glass. Well-ground rims ensure a tight fit.
The diameter of the opening in the top is 2.5 cm.
Size: 13 x 23 cm
Capacity: 2 L
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty