Would you like to log in again?
Day’s Pinchcock Tubing Clamp
Supplier: HUMBOLDT MFG CO
H-8630
470104-108EA 5.35 USD InStock
470104-108
Day’s Pinchcock Tubing Clamp
Clamps Tubing and Sanitary Clamps
A simple wire clamp that may be installed or removed without disconnecting tubing ends.
Made from nickel-plated steel wire with an overall length of 66 mm (2 5/8"). For tubing up to 9.5 mm O.D.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty