Would you like to log in again?
Ward's® Mineral Research Grade Collection
Supplier: VWR
Ward's®
WARD470104-186 WARD470104-400 WARD470104-114 WARD470104-204 WARD470104-240 WARD470104-200 WARD470026-562 WARD470026-560 WARD470104-494 WARD470104-184 WARD470104-416 WARD470104-048 WARD470104-418 WARD470104-206 WARD470104-116 WARD470104-344 WARD470104-292 WARD470104-486 WARD470104-286 WARD470178-584 WARD470104-572 WARD470178-590 WARD470350-962 WARD470026-590 WARD470104-404 WARD470104-424 WARD470026-584 WARD470104-294 WARD470104-118 WARD470104-508 WARD470104-148 WARD470104-228 WARD470104-100 WARD470104-288 WARD470104-308
470104-292EA 48.09 USD InStock
470104-292 470104-572 470026-562 470104-494 470178-590 470026-584 470026-560 470104-294 470104-114 470104-118 470104-116 470104-418 470350-962 470104-416 470178-584 470026-590 470104-286 470104-186 470104-240 470104-184 470104-400 470104-048 470104-344 470104-200 470104-486 470104-288 470104-100 470104-206 470104-404 470104-228 470104-204 470104-424 470104-148 470104-308 470104-508
Ward's® Mineral Research Grade Collection
Specimens Mineral Specimens Classroom Minerals
Ward's® highest quality and purest mineral offerings.
  • Hand picked by Ward's Geology for purity
  • Intended for microscopic study
  • 10 gram packages
  • Average grain size is 3.75 mm
  • 85% purity rate
Ward's mineral research grade collection provides the highest quality and purest mineral offering. A Ward's Geologist hand selects 10 pure grams of material under the micropscope. The specimens are research grade quality to perform accurate experiments. They are also an excellent teaching tool to view microscopic mineralogical features.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty