Would you like to log in again?
Solar Cell
Supplier: SOLAR MADE SE
3-400
470104-492EA 16.05 USD InStock
470104-492
Solar Cell
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Show How Direct/Indirect Sunlight Provides Power
A clear acrylic case with molded-in lenses holds this silicon solar wafer. The cell is color-coded and comes complete with wire leads (10 cm). Output in full sunlight is up to 400 mA at 0.45 VDC.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty