Would you like to log in again?
Wood Test Tube Rack
Supplier: BENNETT WOOD SPECIALTIES SE
6327900
470104-542EA 13.49 USD InStock
470104-542
Wood Test Tube Rack
Racks Tube Racks
One-piece support is able to accommodate larger size tubes. It holds six 20mm tubes. Size: 12 x 6 x 4.5cm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty