Would you like to log in again?
Rapitest Soil Test Kit
Supplier: LUSTER LEAF PRODUCTS INC SE
1612 1601
470104-846CS 98.95 USD InStock
470104-846 470175-090
Rapitest Soil Test Kit
Standards Environmental Analysis Standards
Perform qualitative soil tests without the use of chemical reagents and test tubes.
  • No hazardous compounds required for testing
  • Colorimetric determination of soil components
  • Instructions included
This kit lets students use a simple three-step process involving water, tablets and color comparators. To test soil, simply mix it with water and let it settle. Then pour the liquid off into the color comparator, add a tablet and compare the liquid with the color film chart for a test reading.
470104-846 includes 10 pH soil test kits and fully instructions.
470175-090 includes 10 tests each for pH, nitrogen, phosphorus and potash and full instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty