Would you like to log in again?
Triangular Supports with Rods
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TSR636 TSR420 TSR524
470105-202EA 44.95 USD InStock
470105-202 470105-204 470105-200
Triangular Supports with Rods
Support Stands
Heavy cast-iron triangular bases.
  • Low flat legs on triangular supports provide excellent stability
Black enamel finish is extremely resistant to wear.
Delivery information: Includes a removable, zinc-plated steel rod.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty