Would you like to log in again?
Adjustable Phase Wave Generator
Supplier: THE SCIENCE SOURCE SE
15445
470105-540EA 213.7 USD InStock
470105-540
Adjustable Phase Wave Generator
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
Produces circular waves from either one or two point sources. The phase relationship is continuously adjustable. A variable, low voltage power supply, such as our 64250-10, allows the speed to be adjusted also. Securely mounted in a molded housing with four non-skid, shock-absorbing feet.
  • Includes two point sources for generating circular waves, and ripple bar for generating straight waves
  • Includes instructions
  • Phase control, with variable amplitude and frequency, for varying interference patterns
Ordering information: Requires a variable low voltage power supply: 1.5 to 3 V, not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty