Would you like to log in again?
Breadboard Jumpers
Supplier: GLOBAL SPECIALTIES SE
WK-1
470105-634EA 26.99 USD InStock
470105-634
Breadboard Jumpers
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Use this colorful, 350 piece assortment of 22 gauge solid wire jumpers to make connections on your breadboards. The cut and formed jumpers range from 0.1" to 5.0" in 14 different lengths in a storage case.
The flexible jumpers are ideal for breadboard use. The wire is flexible, but the ends are solid, making breadboarding quick and easy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty