Would you like to log in again?
Refraction Blocks
Supplier: United Scientific Supplies
RCB075 RCB100
470105-730EA 16.9 USD InStock
470105-730 470105-728
Refraction Blocks
Educational Materials Physics Educational Materials Optics Learning Activities
Rectangular blocks in a variety of sizes and are designed for use as an index of refraction plate.
  • Quality Lucite block
  • Optically clear sides
This Lucite block allows you to investigate refraction and internal reflection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty