Would you like to log in again?
Ward's® Live Mealworm Larvae, Pupae and Beetles (Tenebrio)
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470176-238 WARD470176-790 WARD470180-368 WARD470176-754 WARD470180-370 WARD470176-752 WARD470105-824 WARD470313-152 WARD470180-768 WARD470176-792
470176-790PK 35.95 USD InStock
470176-790 470180-368 470105-824 470176-792 470180-768 470180-370 470176-752 470176-238 470176-754 470313-152
Ward's® Live Mealworm Larvae, Pupae and Beetles (Tenebrio)
Specimens Live Specimens Animals - Living Material
Teach your students about the complete metamorphosis of an insect with these mealworms.
  • Store mealworms in bran until ready to use
  • Excellent food source for turtles, salamanders and fish
  • Demonstrates complete metamorphosis
Ordering information: Culture Kit includes culture pan, tenebrio nutrient, and a set of mixed stages as a starter culture.
Delivery information: These products contain living materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Packaging: Larvae and beetle cultures come packed with some tenebrio nutrient.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty