Would you like to log in again?
Snap Circuits Educational Electronics Training Programs
Supplier: ELENCO ELECTRONICS INC SE
SC300R SC-100R-A
470105-896KT 102.6 USD InStock
470105-896 470007-232
Snap Circuits Educational Electronics Training Programs
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The award-winning Snap Circuit Series uses building blocks with snaps to build electrical and electronic circuit projects.
  • Experiment Manual Included
  • Five Hundred Experiments Included
  • Storage Box Included
  • Experiment Topics Include Basic Components and Circuits, Capacitors, and More
  • Series Uses Building Blocks with Snaps to Build Electrical Projects
Each block has a function, and the blocks are color coded and numbered so they're easy to identify.
The blocks feature 500 experiments including basic components and circuits, motors and electricity, resistance, capacitors, transistors, oscillators and electronic sound, integrated circuits, electromagnetism, and radio. Other experiments cover meters, transformers, and FM radio; diodes and applications; and electronic switches.
Ordering information: This kit includes an experiment manual, student guide, and parts organizer/storage box. It requires two AA batteries (not included).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty