Would you like to log in again?
How Clean is your Lab Station? Microbiology Experiment Kit
Supplier: CULTURE MEDIA &SUPPLIES INC SE
67310-05
470105-940EA 61.5 USD InStock
470105-940
How Clean is your Lab Station? Microbiology Experiment Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Test for the presence of bacteria in your workspace before and after cleaning with this TSA Microbiology Experiment Kit.
  • Four popular experiments
Using the included materials, students can conduct a variety of bacteria tests to discover the characteristics of bacteria and conditions under which it is most prevalent.
The unique set up of this kit allows students to melt the media in a microwave, pour their own media plates, and test for bacteria growth in four popular experiments.
Kit Includes:
Two 200ml bottles of prepared Tryptic Soy Agar
20 gamma radiated sterile Petri dishes
10 sterile swabs
Instruction sheets.
Delivery information: This item will ship immediately upon order, unless you specify otherwise.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty