Would you like to log in again?
Refraction Tank with Laser
Supplier: AMERICAN SCIENTIFIC LLC SE
6330-00
470105-996EA 100.15 USD InStock
470105-996
Refraction Tank with Laser
Educational Materials Physics Educational Materials Optics Learning Activities
The refraction tank with laser provides excellent visibility!
  • Rotating laser for easy positioning
  • Demonstrate refraction and internal reflection
Use this sturdy, self-contained device to measure the refraction and incidence of light as well as the critical angle and total internal reflection. Simply fill the circular acrylic tank halfway with water and use the built-in laser, which can be rotated 360° around the tank, to direct a bright, narrow ray of light onto the water surface. The base contains 4 leveling screws to level the water in the tank. Appropriate for Grades 6-12. Requires (1) LR44 button battery (included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty