Would you like to log in again?
Push Ball Soil Thermometer
Supplier: SCIENCE INTERACTIVE GROUP
624-5110
470106-006EA 13.5 USD InStock
470106-006
Push Ball Soil Thermometer
Thermometers Analog Thermometers
Seeds germinate successfully when the soil is the right temperature. This virtually indestructible thermometer gives readings of -15 to 65°C with easy-to-read numbers.
Thermometer includes a round push ball on the top for easy insertion into the soil, even if it is hard or dry. Size: 32 cm long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty