Would you like to log in again?
Boreal Science Quieting Solution
Supplier: ALDON CORP SE
BB0161-30M BB0161-100
470106-046EA 9.25 USD InStock
470106-046 470007-320
Boreal Science Quieting Solution
Chemicals for Science Education
Use this solution to slow down and relax microorganisms such as Paramecium, Euglena and Stentor. The ready-to-use dropper bottle makes it easy to store and transfer quieting liquid. Includes a dropper.
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty