Would you like to log in again?
Quincke’s Interference Tubes
Supplier: GSC INTERNATIONAL SE
904
470106-466EA 49.55 USD InStock
470106-466
Quincke’s Interference Tubes
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
To Accompany Your Tuning Fork!
  • Comes with instructions and suggestions for use.
  • Tuning fork is not included.
This apparatus consists of two Y-tubes about 15 mm in diameter, connected with rubber tubing, with one path being approximately one-half wavelength longer than the other. When a C fork (256 vps) is held near one orifice and the path difference is 65 cm, there will be no sound at the other orifice. When either tube is pinched shut, and the tuning fork is sounded at the intake, sound comes through.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty