Would you like to log in again?
In Search of the Kissing Disease Kit
Supplier: Edvotek
274
470107-680KT 177.4 USD InStock
470107-680
In Search of the Kissing Disease Kit
Educational Materials Biology Educational Materials AP Biology Learning Activities
Apply Biotechnology to Infectious Disease Investigations
  • Convenient Ready-To-Load samples
  • Connection to infectious disease
  • Inquiry-based experiment
  • Time required: ~50 minutes
Infectious mononucleosis is commonly known as the ""kissing disease"". The causative agent is Epstein-Barr virus (EBV) which can be transmitted through saliva during kissing. In this experiment, students search for the presence of EBV using the ELISA reaction to detect specific viral proteins.
Kit includes: instructions, samples, antigens; antibodies, various solutions and reagents, pipettes and microtest tubes.
Required, but not included: 37°C incubation oven, micropipettes with tips, laboratory glassware, distilled or deionized water.
Packaging: Store Ready-to-Load QuickStrip™ samples in the refrigerator
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty