Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Student Lab Tables for 2 Students, 0.75" Phenolic Top

P7204 P7604 P7104 P7304K30N
470107-754EA 1400 USD InStock
470107-754 470107-756 470107-750 470107-752
Student Lab Tables for 2 Students, 0.75" Phenolic Top
Furniture Tables and Workbenches
Legs and aprons are constructed of solid red oak and with heavy-duty 13-gauge steel corner braces. They are smoothly machined with a chemical resistant, earth-friendly UV finish.

  • 2.25" solid square legs, with ⁵/₁₆" hanger bolts installed, supplied nuts and washers
  • 0.75" phenolic work surface
  • Equipped with non-skid adjustable glide for precise leveling and for prevention of rocking on uneven floors
  • Black rubber leg boots which offer protection against scuffing and chemical spills
  • Standard table height is 30" with a weight capacity of 500 lbs.

Color: Northwood oak/black

Minimal assembly required, legs need to be atttached only.

Certifications: SEFA compliant and MAS Certified Green.

Packaging: Shipped KD with tops attached.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty