Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Function Generator

Supplier: B & K PRECISION SE
4010A
470110-206EA 467.99 USD InStock
470110-206
Function Generator
Batteries
Our most economical full feature model.

  • Frequency range 0.2 Hz to 2 MHz in 7 steps
  • Dial accuracy: ±5%
  • Separate adjustments for output level, DC effect, CMOS level and duty cycle
  • VCG input for sweep operation (0-10 V DC input causes 100:1 frequency change)
  • 20 dB step attenuator
  • Output impedance: 50 Ω ±10%

This function generator produces sine, square, triangle, pulse, and ramp signals and has a separate TTL/CMOS output. Comes with an output cable (BNC/insulated clips) and an instruction manual.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty