Would you like to log in again?
Gravitational Balance
Supplier: United Scientific Supplies
CGB001 CGBWR-PK/5
470110-380EA 1686.05 USD InStock
470110-380 470158-410
Gravitational Balance
Educational Materials Physics Educational Materials Mechanics Learning Activities
Simple, But Amazingly Sensitive - Measure One of the Universe's Fundamental Constants!
  • Demonstrates the existence of gravitational forces between masses
  • Measures the Universal Gravitational Constant, G
  • Verifies the Inverse Square Law for gravitational forces
This innovative apparatus, developed from the traditional Cavendish pattern, is substantially less expensive than earlier models, making the measurement of the tiny but highly significant Gravitational Constant more accessible to students. It is precisely constructed and allows measurements of G to be made to better than 15% relative error using either initial acceleration method or the equilibrium displacement method derives G from the change in the equilibrium position of the balance at rest due to the change in position of the large masses. The builit-in oil damping system greatly reduces the time needed for equilibrium form the 2 hours needed by an undamped system.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty