Would you like to log in again?
Electrolytic Bridge
Supplier: LD DIDACTIC GMBH
667 455
470110-430EA 164.9 USD InStock
470110-430
Electrolytic Bridge
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Students measure the voltages generated by a series of different pairs of half cells using the Electrolytic Bridge.
  • Y Tube Length: 9 cm
  • Bridge Manipulates Half Cells so Their Voltages Can Be Measured
  • Ceramic Material Lets Ions Pass but Stops Electrolytic Fluid
  • Students Measure Voltages Created by Pairs of Half Cells
When you immerse an electrode in its own salt, a half-cell voltage is generated between the electrode and the solution. Measuring half-cell voltages is difficult as they have only one electrode. The Electrolytic Bridge separates the cell into two halves and connects them with a separate electrolyte. By measuring a series of electrode materials against a single standard, half-cell voltages can be established.
The apparatus consists of a Y tube with an overall length of 9 cm. The end of each arm is fitted with a special ceramic material that lets ions pass but filters electrolytic fluid.
Ordering information: This kit requires a voltmeter, sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty